Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
4-Methyl-2-pentanone ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur. for extraction analysis, Riedel-de Haën™
 
undefined
 
 
4-Methyl-2-pentanone ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur. for extraction analysis, Riedel-de Haën™
 

 

  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 10 ppm
  • Water (Karl Fischer):
    Max. 500 ppm
  • Titrable Acid:
    Max. 0.002 meq/g
  • Aluminium (Al):
    Max. 0.5 ppm
  • Boron (B):
    Max. 0.02 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Tin (Sn):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • APHA:
    Max. 15
  • Appearance:
    complying
  • Explosion Limit:
    1.2-8 %, 93 °F

 

Frequently Bought Together