Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
 
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
 
 
 
 
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™

 

  • Assay (GC):
    Min. 99.5 %
  • Boiling range:
    123 - 126 °C
  • Explosion Limit:
    24.5 %
  • Density (D 20/20):
    0.966 - 0.968
  • Refractive index (n 20/D):
    1.4010 - 1.4030
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Titrable Acid:
    Max. 0.002 meq/g
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00004 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.0004 %
  • Acetaldehyde:
    Max. 0.001 %
  • Formaldehyde:
    Max. 0.001 %
  • Peroxides (as H₂O₂):
    Max. 0.0003 %
  • APHA:
    Max. 10

 

Frequently Bought Together