Dichloromethane ≥99.9% (by GC) stabilized, Puriss. p.a. ACS, ISO, Riedel-de Haën™
:75-09-2
:
:CH₂Cl₂
:MFCD00000881
:84.93 g/mol
:39…41 °C (1.013 hPa)
:–97 °C
:1.32…1.33 g/cm³ (20 °C)
:Ambient
:200-838-9
:1593
:6.1,III
:GHS07
:GHS08
- Vapor Pressure:24.45 psi ( 55 °C)
- Assay (GC):Min. 99.9 %
- Boiling range:39.5 - 40.5 °C
- Density (D 20/4):1.324 - 1.326
- Refractive index (n 20/D):1.4235 - 1.4245
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as HCl):Max. 0.001 %
- Titrable Acid:Max. 0.0003 meq/g
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Potassium (K):Max. 0.00005 %
- Lithium (Li):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Molybdenum (Mo):Max. 0.00001 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Strontium (Sr):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- Free chlorine (Cl):Max. 0.00002 %
- Free halogens:complying
- Chloride (Cl):Max. 0.00005 %
- Ethanol (GC):Max. 0.05 %
- Formaldehyde:Max. 0.0005 %
- 2-methyl-2-buten (GC):10 - 60 ppm
- Tetrachloromethane (GC):Max. 0.01 %
- Trichloromethane (GC):Max. 0.01 %
- Reaction against H₂SO₄:complying
- APHA:Max. 10
- Autoignition Temperature:1223 °F