

Methanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Boiling Pt:64…65 °C (1.013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
:GHS08
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 Solvent selection guide recommended after discussion
Specifications
Specification Test Results
- Appearance (form):complying
- Appearance (color):complying
- Appearance:complying
- Assay (GC):Min. 99.8 %
- Boiling range:64 - 65 °C
- Density (D 20/20):0.791 - 0.793
- Refractive index (n 20/D):1.328 - 1.330
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Free acid (as HCOOH):Max. 0.002 %
- Free alkali (as NH₃):Max. 0.00005 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000001 %
- Iron (Fe):Max. 0.00001 %
- Potassium (K):Max. 0.00005 %
- Lithium (Li):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000001 %
- Molybdenum (Mo):Max. 0.00001 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.000002 %
- Tin (Sn):Max. 0.00001 %
- Strontium (Sr):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- Chloride (Cl):Max. 0.00005 %
- Sulfate (SO₄):Max. 0.0001 %
- KMnO₄ reducing matter (as O):Max. 0.00025 %
- Acetone (GC):Max. 0.001 %
- Ketones (as CH₃COCH₃):Max. 0.001 %
- Aldehydes (as CH₃CHO):Max. 0.001 %
- Acetaldehyde:Max. 0.001 %
- Ethanol (GC):Max. 0.1 %
- Formaldehyde:Max. 0.001 %
- Carbonyl compounds (as CO):Max. 0.005 %
- Carbonyl compounds (ACS):complying
- Mixable with H₂O:complying
- Reaction against H₂SO₄:complying
- APHA:Max. 10