

Iodine 0.5 M I2 (1.0N) volumetric solution, Fluka™
Catalog # BJ319007-500ML
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:500 ml
- Cat. No.:BJ319007-500ML
Specifications
Specification Test Results
- Assay:0.950 - 1.050 N
Specification Test Results
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Melting Pt:no data available
Density:1.32 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
:GHS05
:GHS08