
Soda lime, granular ACS +100 mesh, Fluka™
Catalog # BJ266434-500G
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:500 g
- Cat. No.:BJ266434-500G
Specifications
Specification Test Results
- Form:granular
- Loss on drying (200°C):Max. 7 %
- Grain size < 100 mesh:Max. 1 %
- CO₂-absorption capacity:Min. 19.0 %
- Capacity:≥19.0 % absorption capacity (CO₂)
- Particle Size:+100 mesh
Specification Test Results
About this item
CAS Number:8006-28-8
:
Storage Temperature:Ambient
UN:1907
ADR:8,III
:GHS05
MDL Number:MFCD00134094