

tert-Butanol ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:75-65-0
:
Formula:(CH₃)₃COH
MDL Number:MFCD00004464
:74.12 g/mol
Melting Pt:25 °C
Storage Temperature:Ambient
EINECS:200-889-7
UN:1120
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- Identity (IR):complying
- Identity (NMR):complying
- Assay (GC):Min. 99.7 %
- Solidification Point:24.0 - 25.5 °C
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.1 %
- Titrable Acid:Max. 0.001 meq/g
- Aluminium (Al):Max. 0.5 ppm
- Boron (B):Max. 0.02 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Tin (Sn):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- Carbonyl Compounds (as HCHO):Max. 100 ppm
- APHA:Max. 20
- Vapor Pressure:31 mmHg ( 20 °C)
- Refractive Index:n20/D 1.387 (lit.)