


Xylene (mixture of isomers) ≥99% (isomers plus ethylbenzene)
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Boiling Pt:136…140 °C
Melting Pt:–34 °C
Density:0.87 g/cm³
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:215-535-7
UN:1307
ADR:3,III
:GHS07
:GHS08
:GHS02
Specification Test Results
- Assay Xylene isomers and Ethylbenzol (GC):Min. 99.0 %
- Ethyl benzene (GC):Max. 25.0 %
- Non-volatile matter:Max. 0.002 %
- IR spectrum:complying
- Refractive Index:n20/D 1.497 (lit.)
- Vapor Pressure:18 mmHg ( 37.7 °C)