

Ammonia solution ≥25% (NH₃ basis) in water, TraceSELECT™ for trace analysis, Fluka™
Catalog # BJ09857-500ML
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:500 ml
- Cat. No.:BJ09857-500ML
Specifications
Specification Test Results
- Assay of NH₃:25.0 - 31.0 %
- Density (D 20/4):0.890 - 0.910
- Silver (Ag):Max. 0.5 ppb
- Aluminium (Al):Max. 5 ppb
- Arsenic (As):Max. 1 ppb
- Gold (Au):Max. 0.5 ppb
- Barium (Ba):Max. 0.5 ppb
- Beryllium (Be):Max. 0.5 ppb
- Bismuth (Bi):Max. 0.5 ppb
- Calcium (Ca):Max. 10 ppb
- Cadmium (Cd):Max. 0.5 ppb
- Cerium (Ce):Max. 0.5 ppb
- Cobalt (Co):Max. 0.5 ppb
- Chromium (Cr):Max. 1 ppb
- Cesium (Cs):Max. 0.5 ppb
- Copper (Cu):Max. 0.5 ppb
- Iron (Fe):Max. 2 ppb
- Gallium (Ga):Max. 0.5 ppb
- Germanium (Ge):Max. 0.5 ppb
- Mercury (Hg):Max. 1 ppb
- Indium (In):Max. 0.5 ppb
- Iridium (Ir):Max. 0.5 ppb
- Potassium (K):Max. 5 ppb
- Lithium (Li):Max. 0.5 ppb
- Magnesium (Mg):Max. 1 ppb
- Manganese (Mn):Max. 0.5 ppb
- Molybdenum (Mo):Max. 0.5 ppb
- Sodium (Na):Max. 10 ppb
- Nickel (Ni):Max. 1 ppb
- Lead (Pb):Max. 0.5 ppb
- Palladium (Pd):Max. 0.5 ppb
- Platinum (Pt):Max. 0.5 ppb
- Rubidium (Rb):Max. 0.5 ppb
- Rhodium (Rh):Max. 0.5 ppb
- Ruthenium (Ru):Max. 0.5 ppb
- Antimony (Sb):Max. 0.5 ppb
- Selenium (Se):Max. 1 ppb
- Tin (Sn):Max. 0.5 ppb
- Strontium (Sr):Max. 0.5 ppb
- Titanium (Ti):Max. 1 ppb
- Thallium (Tl):Max. 0.5 ppb
- Uran (U):Max. 0.5 ppb
- Vanadium (V):Max. 1 ppb
- Zinc (Zn):Max. 1 ppb
- Zirconium (Zr):Max. 0.5 ppb
- Carbonate (as CO₂):Max. 10 ppm
- Bromide (Br):Max. 50 ppb
- Chloride (Cl):Max. 500 ppb
- Fluoride (F):Max. 100 ppb
- Iodide (I):Max. 50 ppb
- Nitrite (NO₂):Max. 100 ppb
- Nitrate (NO₃):Max. 500 ppb
- Phosphate (PO₄):Max. 50 ppb
- Sulfate (SO₄):Max. 100 ppb
Specification Test Results
About this item
CAS Number:1336-21-6
:
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:37.7 °C (1.013 hPa)
Melting Pt:–92 °C
Density:0.90 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2672
ADR:8,III
:GHS07
:GHS05
:7664-41-7