Order Entry
Puerto Rico
ContactUsLinkComponent
Simian virus V5 Epitope Tag
Simian virus V5 Epitope Tag
Catalog # 103006-384
Supplier:  Anaspec
CAS Number:  
Simian virus V5 Epitope Tag
Catalog # 103006-384
Supplier:  Anaspec
Supplier Number:  AS-61176
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Simian virus
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Simian virus 5 epitope tag|SV5 tag
  • Protein/Peptide Name:
    V5 Epitope Tag
  • Purity:
    95%
  • Molecular Weight:
    1421.7 Da
  • Sequence:
    GKPIPNPLLGLDST
  • Cat. No.:
    103006-384
  • Supplier no.:
    AS-61176

Specifications

About this item

This sequence is from the C-terminal sequence of the P and V proteins of Simian Virus 5.
Sequence:GKPIPNPLLGLDST
MW:1421.7 Da
% peak area by HPLC:95
Storage condition:-20° C