Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:Transdermal protein delivery Peptide
- Protein/Peptide Name:Transdermal Peptide
- Purity:95%
- Molecular Weight:1063.2 Da
- Sequence:ACSSSPSKHCG
- Cat. No.:103007-172
- Supplier no.:AS-62066
This short synthetic peptide facilitates efficient transdermal protein drug delivery through intact skin. Co-administration of the peptide and insulin to the abdominal skin of diabetic rats results in elevated systemic levels of insulin and suppresses serum glucose levels. This peptide creates a transient opening in the skin barrier to enable macromolecular drugs to reach systemic circulation.
Sequence:ACSSSPSKHCG
MW:1063.2 Da
% peak area by HPLC:95
Storage condition:-20° C