Order Entry
Puerto Rico
ContactUsLinkComponent
Pep-1-Cysteamine
Pep-1-Cysteamine
Catalog # 103007-778
Supplier:  Anaspec Inc
CAS Number:  
Pep-1-Cysteamine
Catalog # 103007-778
Supplier:  Anaspec Inc
Supplier Number:  AS-63849
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    CPP|Cell penetrating Peptide
  • Protein/Peptide Name:
    Pep-1-Cysteamine
  • Purity:
    95%
  • Molecular Weight:
    2950.3 Da
  • Sequence:
    Ac-KETWWETWWTEWSQPKKKRKV-cysteamine
  • Cat. No.:
    103007-778
  • Supplier no.:
    AS-63849

Specifications

About this item

Pep-1 is one of the synthetic cell-penetrating peptides (CPPs), which has been successfully used to deliver a variety of proteins and other biopharmaceutical macromolecules into cells in a non-disruptive way. It is a CPP with primary amphipathicity (i.e amphipathicity resulting from the amino acid sequence itself, not from the folding structure) that comprises a tryptophan-rich so-called ‘hydrophobic’ domain, a hydrophilic domain derived from an NLS (nuclear localization signal) of SV40 (simian virus 40) large T-antigen, and a spacer between them. A cysteamine group is present at the C-terminus. The presence of cysteamine group in C terminal seems to play a crucial role in the delivery efficiency of cargoes into cells.
Sequence:Ac-KETWWETWWTEWSQPKKKRKV-cysteamine
MW:2950.3 Da
% peak area by HPLC:95
Storage condition:-20° C