Order Entry
Puerto Rico
ContactUsLinkComponent
Penetratin-Arg
Penetratin-Arg
Catalog # 103009-970
Supplier:  Anaspec Inc
CAS Number:  
Penetratin-Arg
Catalog # 103009-970
Supplier:  Anaspec Inc
Supplier Number:  AS-65464
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Penetratin Arginine Peptide
  • Protein/Peptide Name:
    Penetratin-Arg
  • Purity:
    95%
  • Molecular Weight:
    2358.8 Da
  • Sequence:
    RQIRIWFQNRRMRWRR
  • Cat. No.:
    103009-970
  • Supplier no.:
    AS-65464

Specifications

About this item

Penetratin-Arg is a cell-penetrating peptide (CPP) that is derived from the 3rd helix of Drosophila Antennapedia homeodomain protein. Penetratin-Arg is the same as Penetratin except that Lysine residues were substituted with Arginines. It forms an alpha-helix structure in lipid environment and can permeate cell membrane at low micromolar concentration without significantly affecting membrane. CPP can be conjugated with large molecules and used as a drug delivery vehicle. R
Sequence:RQIRIWFQNRRMRWRR
MW:2358.8 Da
% peak area by HPLC:95
Storage condition:-20° C