Order Entry
Puerto Rico
ContactUsLinkComponent
(Arg)9, AnaSpec
(Arg)9, AnaSpec
Catalog # 103006-406
Supplier:  Anaspec
CAS Number:  
(Arg)9, AnaSpec
Catalog # 103006-406
Supplier:  Anaspec
Supplier Number:  AS-61204
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Cell permeable Peptide|Arginine repeat|CPP
  • Protein/Peptide Name:
    (Arg)9
  • Purity:
    95%
  • Molecular Weight:
    1423.7 Da
  • Sequence:
    RRRRRRRRR
  • Cat. No.:
    103006-406
  • Supplier no.:
    AS-61204

Specifications

About this item

(Arg)9 is a cell-permeable peptide used for drug delivery.It can traverse the plasma membrane of eukaryotic cells.
Sequence:RRRRRRRRR
MW:1423.7 Da
% peak area by HPLC:95
Storage condition:-20° C