Order Entry
Puerto Rico
ContactUsLinkComponent
Interphotoreceptor Retinoid Binding Protein Fragment
Interphotoreceptor Retinoid Binding Protein Fragment
Supplier:  Anaspec Inc
Certificates
Interphotoreceptor Retinoid Binding Protein Fragment
Supplier:  Anaspec Inc

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    IRBP Peptide|T-cell epitope (161-180)
  • Protein/Peptide Name:
    Interphotoreceptor Retinoid Binding Protein Fragment
  • Purity:
    95%
  • Molecular Weight:
    2209.5 Da
  • Sequence:
    SGIPYIISYLHPGNTILHVD

Specifications

Related Information