Order Entry
Puerto Rico
ContactUsLinkComponent
Saccharomyces cerevisiae alpha-Mating Factor Pheromone
Saccharomyces cerevisiae alpha-Mating Factor Pheromone
Catalog # 103003-004
Supplier:  Anaspec
CAS Number:  
Saccharomyces cerevisiae alpha-Mating Factor Pheromone
Catalog # 103003-004
Supplier:  Anaspec
Supplier Number:  AS-60221-5
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Saccharomyces cerevisiae
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    alpha factor pheromone Peptide
  • Protein/Peptide Name:
    alpha-Mating Factor Pheromone
  • Purity:
    95%
  • Molecular Weight:
    1684 Da
  • Sequence:
    WHWLQLKPGQPMY
  • Cat. No.:
    103003-004
  • Supplier no.:
    AS-60221-5

Specifications

About this item

This tridecapeptide, an alpha-factor pheromone of Saccharomyces cerevisiae, induces conjugation in yeast by binding to Ste2p. Its cognate GPCR activates a G protein signal pathway that highly conserves with mammalian signaling pathways.
Sequence:WHWLQLKPGQPMY
MW:1684 Da
% peak area by HPLC:95
Storage condition:-20° C