Order Entry
Puerto Rico
ContactUsLinkComponent
 
Cathepsin S Substrate, AMC (7-amino-4-methylcoumarin)
  103007-070
 :  
Cathepsin S Substrate, AMC (7-amino-4-methylcoumarin)
  103007-070
 :  AS-61859
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    AMC (7-amino-4-methylcoumarin)
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Cathepsin FRET Peptide|Cathepsin S
  • Protein/Peptide Name:
    Cathepsin S Substrate
  • Purity:
    95%
  • Molecular Weight:
    871.1 Da
  • Sequence:
    Ac-KQKLR-AMC
  • Cat. No.:
    103007-070
  • Supplier no.:
    AS-61859

 

 

Cathepsins are a class of globular lysosomal proteases, playing a vital role in mammalian cellular turnover. They degrade polypeptides and are distinguished by their substrate specificities. Cathepsin S is a cysteine proteinase involved in the pathogenesis of autoimmune diseases, atherosclerosis, cancer, obesity and related diseases.
This peptide is a cathepsin S substrate fluorescently labeled with AMC (Ex/Em=354 nm/442 nm). It can be used to measure cathepsin S activity.
Sequence:Ac-KQKLR-AMC
MW:871.1 Da
% peak area by HPLC:95
Storage condition:-20° C