Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Multiple Antigenic Peptide

Multiple Antigenic Peptide

10 mg
Catalog # 102998-700
Marketsource
Supplier:   Anaspec
Supplier #:  AS-21006
Multiple Antigenic Peptide
Catalog # 102998-700
Supplier:  Anaspec
Marketsource
CAS Number:  
Multiple Antigenic Peptide

10 mg

CAS Number:  
Price
$477.92
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Multiple Antigenic Peptides (MAPs) are synthetic peptides with 4 branches harboring 2 Lysines each. The Lys residues are used as a scaffold to support 8 peptides. MAPs are used to increase the mass of the peptide entity when used as antigen in immunizations. They represent an alternative to carrier proteins, such as BSA or KLH.
Sequence:K4K2KA - NH2
MW:985.3 Da
% peak area by HPLC:95
Storage condition:-20° C

Documents

Specifications

Specifications

  • Size:
    10 mg
  • Storage Conditions:
    –20 °C
  • Cat. No.:
    102998-700
  • Molecular Weight:
    985.3 Da
  • Purity:
    95%
  • Protein Synonyms:
    Multiple antigenic Peptides
  • Protein/Peptide Name:
    Multiple Antigenic Peptide
  • Conjugation:
    Unconjugated
  • Sequence:
    K4K2KA - NH2

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.