Order Entry
Puerto Rico
ContactUsLinkComponent
epsilon-V1-2
epsilon-V1-2
Catalog # 103007-228
Supplier:  Anaspec Inc
CAS Number:  
epsilon-V1-2
Catalog # 103007-228
Supplier:  Anaspec Inc
Supplier Number:  AS-62187
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    PKC inhibitor Peptide
  • Protein/Peptide Name:
    epsilon-V1-2
  • Purity:
    95%
  • Molecular Weight:
    947.1 Da
  • Sequence:
    EAVSLKPTC
  • Cat. No.:
    103007-228
  • Supplier no.:
    AS-62187

Specifications

About this item

This peptide is the εPKC specific inhibitor. Its inhibitory activity is based on εPKC translocation and MARCKS phosphorylation. This peptide interferes with εPKC interaction with the anchoring protein εRACK. This peptide contains a cysteine residue added to the C-terminus for potential S-S bond formation with a carrier protein
Sequence:EAVSLKPTC
MW:947.1 Da
% peak area by HPLC:95
Storage condition:-20° C