Order Entry
Puerto Rico
ContactUsLinkComponent
[Ser25]-PKC (19-31),Biotin
[Ser25]-PKC (19-31),Biotin
Catalog # 102996-368
Supplier:  Anaspec Inc
CAS Number:  
[Ser25]-PKC (19-31),Biotin
Catalog # 102996-368
Supplier:  Anaspec Inc
Supplier Number:  AS-23018
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Biotin
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    PKC pseudosubstrate Peptide|PKC regulatory domain Peptide
  • Protein/Peptide Name:
    [Ser25]-PKC (19-31)
  • Purity:
    95%
  • Molecular Weight:
    1914.3 Da
  • Sequence:
    K(Biotin)-RFARKGSLRQKNV
  • Cat. No.:
    102996-368
  • Supplier no.:
    AS-23018

Specifications

About this item

This is a peptide derived from the pseudosubstrate regulatory domain of PKC α residues (19-31) with alanine being replaced with serine at position 25.
Sequence:K(Biotin)-RFARKGSLRQKNV
MW:1914.3 Da
% peak area by HPLC:95
Storage condition:-20° C