Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Jak3tide

Jak3tide

1 mg
Catalog # 103007-388
Marketsource
Supplier:   Anaspec
Supplier #:  AS-62537
Jak3tide
Catalog # 103007-388
Supplier:  Anaspec
Marketsource
CAS Number:  
Jak3tide

1 mg

CAS Number:  
Price
$191.46
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This peptide is a substrate for Jak3. It may be used used in kinase assays. Jak3tide contains the phosphorylation site at Tyr7.
Sequence:GGEEEEYFELVKKKK
MW:1813 Da
% peak area by HPLC:95
Storage condition:-20° C

Documents

Specifications

Specifications

  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Cat. No.:
    103007-388
  • Molecular Weight:
    1813 Da
  • Purity:
    95%
  • Protein Synonyms:
    tyrosine kinase substrate|Janus kinase 3 substrate
  • Protein/Peptide Name:
    Jak3tide
  • Conjugation:
    Unconjugated
  • Sequence:
    GGEEEEYFELVKKKK

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.