Order Entry
Puerto Rico
ContactUsLinkComponent
IRAK-1 (360-380) Peptide Substrate
IRAK-1 (360-380) Peptide Substrate
Catalog # 103008-602
Supplier:  Anaspec
CAS Number:  
IRAK-1 (360-380) Peptide Substrate
Catalog # 103008-602
Supplier:  Anaspec
Supplier Number:  AS-64959
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Interleukin-1 Receptor-Associated Kinase substrate
  • Protein/Peptide Name:
    IRAK-1 (360-380) Peptide Substrate
  • Purity:
    95%
  • Molecular Weight:
    2285.6 Da
  • Sequence:
    KKARFSRFAGSSPSQSSMVAR
  • Cat. No.:
    103008-602
  • Supplier no.:
    AS-64959

Specifications

About this item

This is a substrate peptide for Interleukin-1 Receptor-Associated Kinase (IRAK) 4, derived from the IRAK-1 activation loop peptide amino acids 360 to 380. This peptide sequence contains Ser-376, one of the primary phospho-acceptor residues of IRAK-1. IRAK-4 phosphorylates IRAK-1 as a key step in regulation of inflammatory signaling pathways.
Sequence:KKARFSRFAGSSPSQSSMVAR
MW:2285.6 Da
% peak area by HPLC:95
Storage condition:-20° C