Order Entry
Puerto Rico
ContactUsLinkComponent
DNA-PK Substrate
DNA-PK Substrate
Catalog # 103005-904
Supplier:  Anaspec
CAS Number:  
DNA-PK Substrate
Catalog # 103005-904
Supplier:  Anaspec
Supplier Number:  AS-60210-5
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    DNA-dependent protein kinase substrate
  • Protein/Peptide Name:
    DNA-PK Substrate
  • Purity:
    95%
  • Molecular Weight:
    1759 Da
  • Sequence:
    EPPLSQEAFADLWKK
  • Cat. No.:
    103005-904
  • Supplier no.:
    AS-60210-5

Specifications

About this item

A substrate for DNA-dependent protein kinase (DNA-PK), phosphorylation. DNA-PK is essential for the repair of DNA double-strand breaks. This peptide corresponding to 11–24 amino acids of human p53 with threonine 18 and serine 20 changed to alanine is used as a substrate for the assay of DNA-PK activity
Sequence:EPPLSQEAFADLWKK
MW:1759 Da
% peak area by HPLC:95
Storage condition:-20° C