Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:5 mg
- Storage Conditions:–20 °C
- Protein Synonyms:PKC substrate|CaM kinase II substrate
- Protein/Peptide Name:Syntide-2
- Purity:95%
- Molecular Weight:1507.9 Da
- Sequence:PLARTLSVAGLPGKK
- Cat. No.:102996-268
- Supplier no.:AS-22552
The native peptide, PLARTLSVAGLPGKK (cat# 22511), is a selective substrate for CaM kinase II and protein kinase C. It is a poor substrate for phosphorylase kinase and is not phosphorylated by myosin light chain kinase. The sequence of PLARTLSVAGLPGKK is homologous to phosphorylation site 2 in glycogen synthase. The relative Vmax/Km ratios of the peptide for different kinases are 100 for CaMK II, 22 for protein kinase C, 2 for phosphorylase kinase, and 0.5 for myosin light chain kinase respectively.
Sequence:PLARTLSVAGLPGKK
MW:1507.9 Da
% peak area by HPLC:95
Storage condition:-20° C