Order Entry
Puerto Rico
ContactUsLinkComponent
AMARA peptide
AMARA peptide
Catalog # 103007-410
Supplier:  Anaspec Inc
CAS Number:  
AMARA peptide
Catalog # 103007-410
Supplier:  Anaspec Inc
Supplier Number:  AS-62596
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    AMPK-related kinase substrate
  • Protein/Peptide Name:
    AMARA Peptide
  • Purity:
    95%
  • Molecular Weight:
    1542.8 Da
  • Sequence:
    AMARAASAAALARRR
  • Cat. No.:
    103007-410
  • Supplier no.:
    AS-62596

Specifications

About this item

AMARA peptide is a minimal substrate for several members of the protein kinases family. It contains the phosphorylation site for AMP-activated Protein Kinase (AMPK). AMARA peptide may be employed in the applications to measure AMPK-related kinase activity.
Sequence:AMARAASAAALARRR
MW:1542.8 Da
% peak area by HPLC:95
Storage condition:-20° C