Order Entry
Puerto Rico
ContactUsLinkComponent
Cyclin-dependent kinase 5 peptide
Cyclin-dependent kinase 5 peptide
Catalog # 103002-946
Supplier:  Anaspec Inc
CAS Number:  
Cyclin-dependent kinase 5 peptide
Catalog # 103002-946
Supplier:  Anaspec Inc
Supplier Number:  AS-60026-1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    CDK5 substrate
  • Protein/Peptide Name:
    Cyclin-dependent kinase 5 Peptide
  • Purity:
    95%
  • Molecular Weight:
    1138.5 Da
  • Sequence:
    PKTPKKAKKL
  • Cat. No.:
    103002-946
  • Supplier no.:
    AS-60026-1

Specifications

About this item

The native peptide PKTPKKAKKL (60026-1) is derived from histone H1 peptide sequence that is docked in the active site of cyclin-dependent kinase 5. It is an effective CDK5 substrate (Km = 40 µM).
Sequence:PKTPKKAKKL
MW:1138.5 Da
% peak area by HPLC:95
Storage condition:-20° C