Order Entry
Puerto Rico
ContactUsLinkComponent
 
Rabbit CK1 Peptide Substrate
Catalog # 103007-734
Supplier:  Anaspec Inc
CAS Number:  
Rabbit CK1 Peptide Substrate
Catalog # 103007-734
Supplier:  Anaspec Inc
Supplier Number:  AS-63797
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Rabbit
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    CK1 substrate|Casein Kinase I substrate
  • Protein/Peptide Name:
    CK1 Peptide Substrate
  • Purity:
    95%
  • Molecular Weight:
    1603.9 Da
  • Sequence:
    KRRRAL-pS-VASLPGL
  • Cat. No.:
    103007-734
  • Supplier no.:
    AS-63797

Specifications

About this item

This peptide sequence is based on rabbit muscle glycogen synthase with Ser7 phosphorylated. It is a peptide substrate for Casein Kinase I (CK1). CK1 phosphorylates Ser10. Ser7 is phosphorylated by PKA in vivo.
Sequence:KRRRAL-pS-VASLPGL
MW:1603.9 Da
% peak area by HPLC:95
Storage condition:-20° C