Order Entry
Puerto Rico
ContactUsLinkComponent
Autocamtide-3-Related Inhibitory Peptide
Autocamtide-3-Related Inhibitory Peptide
Catalog # 103008-590
Supplier:  Anaspec Inc
CAS Number:  
Autocamtide-3-Related Inhibitory Peptide
Catalog # 103008-590
Supplier:  Anaspec Inc
Supplier Number:  AS-64930
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    CaMKII inhibitor substrate|Myristoylated Autocatamide derived Peptide|AC3-1|Myristoylated AC3-1|Calmodulin-Dependent Protein Kinase ll inhibitor substrate
  • Protein/Peptide Name:
    Autocamtide-3-Related Inhibitory Peptide
  • Purity:
    95%
  • Molecular Weight:
    1689.1 Da
  • Sequence:
    Myr-KKALHRQEAVDAL
  • Cat. No.:
    103008-590
  • Supplier no.:
    AS-64930

Specifications

About this item

This is a myristoylated form of Autocamtide-3-Derived Inhibitory Peptide (AC3-I), a highly specific inhibitor of Calmodulin-Dependent Protein Kinase ll (CaMKII) that is resistant to proteolysis. AC3-I is derived from Autocamtide-3, a substrate for CaMKII, with the Thr-9 phosphorylation site substituted with Ala.
Sequence:Myr-KKALHRQEAVDAL
MW:1689.1 Da
% peak area by HPLC:95
Storage condition:-20° C