Order Entry
Puerto Rico
ContactUsLinkComponent
 
Histone H4 (1-23)-GSGSK, Biotin
Catalog # 103008-532
Supplier:  Anaspec Inc
CAS Number:  
Histone H4 (1-23)-GSGSK, Biotin
Catalog # 103008-532
Supplier:  Anaspec Inc
Supplier Number:  AS-65002
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Biotin
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Histone H4 Peptide
  • Protein/Peptide Name:
    Histone H4 (1-23)-GSGSK
  • Purity:
    95%
  • Molecular Weight:
    3003.6 Da
  • Sequence:
    SGRGKGGKGLGKGGAKRHRKVLR-GSGSK(Biotin)
  • Cat. No.:
    103008-532
  • Supplier no.:
    AS-65002

Specifications

About this item

This peptide consists of amino acids 1-23 of histone H4 attached at the C-terminal by a GSGS linker to biotinylated lysine. It is used as a substrate for histone acetyltransferase (HAT) assays. Biotinylation allows the peptide to be captured on streptavidin or agarose beads.
Sequence:SGRGKGGKGLGKGGAKRHRKVLR-GSGSK(Biotin)
MW:3003.6 Da
% peak area by HPLC:95
Storage condition:-20° C