To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Biotin
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:H4K5/8/12/16|Histone H4 Peptide
- Protein/Peptide Name:[Lys(Ac)5/8/12/16]-Histone H4 (1-21)-GGK
- Purity:95%
- Molecular Weight:2728.2 Da
- Sequence:SGRG-K(Ac)-GG-K(Ac)-GLG-K(Ac)-GGA-K(Ac)-RHRKV-GGK(Biotin)
- Cat. No.:103008-528
- Supplier no.:AS-64989-1
Specifications
About this item
This peptide is histone H4 (1-21), acetylated at lysine 5, 8, 12, and 16. It contains a C-terminal GG linker, followed by a biotinylated Lys. In general, histone H4 acetylation is associated with increased DNA replication during mitosis, although different combinations of acetylation at specific sites are selective for specific processes. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:SGRG-K(Ac)-GG-K(Ac)-GLG-K(Ac)-GGA-K(Ac)-RHRKV-GGK(Biotin)
MW:2728.2 Da
% peak area by HPLC:95
Storage condition:-20° C