Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:Histone H4 Peptide|P300/CBP
- Protein/Peptide Name:Histone H4 (1-21) Substrate
- Purity:95%
- Molecular Weight:2091.5 Da
- Sequence:SGRGKGGKGLGKGGAKRHRKV
- Cat. No.:103007-372
- Supplier no.:AS-62499
Specifications
About this item
This sequence is amino acids 1 to 21 fragment of the histone 4. Histone acetyltransferase (HAT) p300/CBP catalyses the acetylation of this N-terminal tail of free histone H4 at Lys5, 8, 12, 16. Lys8 is thought to be the preferred acetylation site in H4. Determinants of interaction of p300 with histone 4 appear to be fully present on H4-21, the N-terminal region of the protein.
Sequence:SGRGKGGKGLGKGGAKRHRKV
MW:2091.5 Da
% peak area by HPLC:95
Storage condition:-20° C