Order Entry
Puerto Rico
ContactUsLinkComponent
[Lys(Ac)12]-Histone H4 (1-20)
[Lys(Ac)12]-Histone H4 (1-20)
Catalog # 103007-664
Supplier:  Anaspec Inc
CAS Number:  
[Lys(Ac)12]-Histone H4 (1-20)
Catalog # 103007-664
Supplier:  Anaspec Inc
Supplier Number:  AS-63685
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Histone H4 Peptide|H4K12
  • Protein/Peptide Name:
    [Lys(Ac)12]-Histone H4 (1-20)
  • Purity:
    95%
  • Molecular Weight:
    2034.4 Da
  • Sequence:
    SGRGKGGKGLG-K(Ac)-GGAKRHRK
  • Cat. No.:
    103007-664
  • Supplier no.:
    AS-63685

Specifications

About this item

This peptide is Histone 4 acetylated at lysine 12. Based on genome-wide localization studies, this peptide has been found to occupy promoter region at the transcription start sites, especially being associated with particular promoters that may drive high levels of mRNA transcripts stored in mature spermatozoa. Such observations have led to assumptions that H4K12Ac may be one of the epigenetic marks for developmentally important genes.
Sequence:SGRGKGGKGLG-K(Ac)-GGAKRHRK
MW:2034.4 Da
% peak area by HPLC:95
Storage condition:-20° C