About this item
This is a Histone 3 peptide trimethylated at lysine 36. It's role has been indicative of defining exons, as certain nucleosome-enriched exons are observed to be enriched in some histone modifications. It is belived that this pattern of modification as seen with this H3K36me3 peptide, influences alternative splicing, perhaps by signaling effector proteins to mark particular exons for inclusion in the final transcript as they exit the RNA Polymerase II complex. It has also been shown in yeast that H3K36me3 gets deposited on histones as they are displaced by RNA polymerase II (RNAPII) during transcription. H3K36me3 then serves as a mark for HDACs to bind and deacetylate the histones.
Sequence:ATKAARKSAPATGGV-K(Me3)-KPHRYRPG-GK(Biotin)
MW:2959.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Biotin
- Storage Conditions:–20 °C
- Protein Synonyms:Histone H3 Peptide|H3K36
- Protein/Peptide Name:[Lys(Me3)36]-Histone H3 (21-44)-GK
- Purity:95%
- Molecular Weight:2959.5 Da
- Sequence:ATKAARKSAPATGGV-K(Me3)-KPHRYRPG-GK(Biotin)