Order Entry
Puerto Rico
ContactUsLinkComponent
 
[Lys(Me2)9]-Histone H3 (1-21)
  103009-506
 :  
[Lys(Me2)9]-Histone H3 (1-21)
  103009-506
 :  AS-65401
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Histone H3 Peptide
  • Protein/Peptide Name:
    [Lys(Me2)9]-Histone H3 (1-21)
  • Purity:
    95%
  • Molecular Weight:
    2282.6 Da
  • Sequence:
    ARTKQTAR-K(Me2)-STGGKAPRKQLA
  • Cat. No.:
    103009-506
  • Supplier no.:
    AS-65401

 

 

This is histone H3 (1-21) dimethylated at Lys9. Methylation of Lys 9 is associated with transcriptionally silent chromatin. Histone H3 mono- and dimethylated at Lys 9 localize specifically to silent domains within euchromatin while histone H3 trimethylated at this residue localizes to pericentric heterochromatin.
Sequence:ARTKQTAR-K(Me2)-STGGKAPRKQLA
MW:2282.6 Da
% peak area by HPLC:95
Storage condition:-20° C