Order Entry
Puerto Rico
ContactUsLinkComponent
[Lys(Me1)9]-Histone H3 (1-21)
[Lys(Me1)9]-Histone H3 (1-21)
Catalog # 103009-504
Supplier:  Anaspec
CAS Number:  
[Lys(Me1)9]-Histone H3 (1-21)
Catalog # 103009-504
Supplier:  Anaspec
Supplier Number:  AS-65398
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Histone H3 Peptide|H3K9
  • Protein/Peptide Name:
    [Lys(Me1)9]-Histone H3 (1-21)
  • Purity:
    95%
  • Molecular Weight:
    2269.1 Da
  • Sequence:
    ARTKQTAR-K(Me1)-STGGKAPRKQLA
  • Cat. No.:
    103009-504
  • Supplier no.:
    AS-65398

Specifications

About this item

This is a monomethylated lysine at position 9 of the 1-21 amino acid histone H3 peptide. This form of histone methylation is characterized as being enriched around transcription start sites. This peptide has been used as a control peptide in a TR-FRET assay for identifying inhibitors of histone lysine demethylases.
Sequence:ARTKQTAR-K(Me1)-STGGKAPRKQLA
MW:2269.1 Da
% peak area by HPLC:95
Storage condition:-20° C