Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:Histone H3 Peptide
- Protein/Peptide Name:[Lys(Me1)4]-Histone H3 (1-21)
- Purity:95%
- Molecular Weight:2268.6 Da
- Sequence:ART-K(Me1)-QTARKSTGGKAPRKQLA
- Cat. No.:103008-186
- Supplier no.:AS-64459
Specifications
About this item
This is a monomethylated lysine at position 4 of the 1-21 amino acid histone H3 peptide. This form of histone methylation is characterized as being enriched around transcription start sites. It’s chromatin mark was also closely correlated to cell-type specific expression of putative gene targets of enhancers. It was also noted from a genome wide study that most potential regulatory elements were enriched only by the mono-methylated version of this histone 3.
Sequence:ART-K(Me1)-QTARKSTGGKAPRKQLA
MW:2268.6 Da
% peak area by HPLC:95
Storage condition:-20° C