Order Entry
Puerto Rico
ContactUsLinkComponent
[Arg(Me2s)8]-Histone H3 (1-21),Biotin
[Arg(Me2s)8]-Histone H3 (1-21),Biotin
Catalog # 103009-906
Supplier:  Anaspec
CAS Number:  
[Arg(Me2s)8]-Histone H3 (1-21),Biotin
Catalog # 103009-906
Supplier:  Anaspec
Supplier Number:  AS-65351-1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Biotin
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Histone H3 Peptide
  • Protein/Peptide Name:
    [Arg(Me2s)8]-Histone H3 (1-21)
  • Purity:
    95%
  • Molecular Weight:
    2637.1 Da
  • Sequence:
    ARTKQTA-R(Me2s)-KSTGGKAPRKQLA-K(Biotin)
  • Cat. No.:
    103009-906
  • Supplier no.:
    AS-65351-1

Specifications

About this item

This is histone H3 (1-21) symmetrically dimethylated at Arg8 with a methyl group added to each nitrogen of the guanidinium group. This peptide is biotinylated at the epsilon side chain of an additional C-terminal Lys. Methylation at Arg8 blocks G9a methylation of Lys9. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ARTKQTA-R(Me2s)-KSTGGKAPRKQLA-K(Biotin)
MW:2637.1 Da
% peak area by HPLC:95
Storage condition:-20° C