Order Entry
Puerto Rico
ContactUsLinkComponent
[Lys(Ac)14]-Histone H3 (1-19)
[Lys(Ac)14]-Histone H3 (1-19)
Catalog # 103007-658
Supplier:  Anaspec Inc
CAS Number:  
[Lys(Ac)14]-Histone H3 (1-19)
Catalog # 103007-658
Supplier:  Anaspec Inc
Supplier Number:  AS-63682
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Histone H3 Peptide|H3K14
  • Protein/Peptide Name:
    [Lys(Ac)14]-Histone H3 (1-19)
  • Purity:
    95%
  • Molecular Weight:
    2112.4 Da
  • Sequence:
    ARTKQTARKSTGG-K(Ac)-APRKQ
  • Cat. No.:
    103007-658
  • Supplier no.:
    AS-63682

Specifications

About this item

This peptide is Histone H3 acetylated at lysine 14. Histone lysine aetylation is known to play an important role in chromatin-directed gene transcription. A bromodomain 2 of polybromo (a chromatin remodelling protein) preferentially recognizes acetylated lysine of H3.
Sequence:ARTKQTARKSTGG-K(Ac)-APRKQ
MW:2112.4 Da
% peak area by HPLC:95
Storage condition:-20° C