Order Entry
Puerto Rico
ContactUsLinkComponent
Nuclear Factor (Erythroid-derived 2)like 2 (74-87)
Nuclear Factor (Erythroid-derived 2)like 2 (74-87)
Catalog # 103009-894
Supplier:  Anaspec
CAS Number:  
Nuclear Factor (Erythroid-derived 2)like 2 (74-87)
Catalog # 103009-894
Supplier:  Anaspec
Supplier Number:  AS-65341
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Neh2 nuclear factor domain Peptide|Nrf2 Peptide
  • Protein/Peptide Name:
    Nuclear Factor (Erythroid-derived 2)like 2 (74-87)
  • Purity:
    95%
  • Molecular Weight:
    1631.8 Da
  • Sequence:
    LQLDEETGEFLPIQ
  • Cat. No.:
    103009-894
  • Supplier no.:
    AS-65341

Specifications

About this item

This peptide is derived from the Neh2 domain of nuclear factor (erythroid-derived 2)-like 2, or Nrf2. Nrf2 is a bZIP transcription factor that regulates the expression of antioxidative and cytoprotective genes. The DxETGE motif of this peptide binds Keap1 Kelch adaptor protein and displaces Nrf2 from Keap1.
Sequence:LQLDEETGEFLPIQ
MW:1631.8 Da
% peak area by HPLC:95
Storage condition:-20° C