Order Entry
Puerto Rico
ContactUsLinkComponent
LyP-1, Peptide 1
LyP-1, Peptide 1
Catalog # 103007-222
Supplier:  Anaspec Inc
CAS Number:  
LyP-1, Peptide 1
Catalog # 103007-222
Supplier:  Anaspec Inc
Supplier Number:  AS-62169
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Lyp-1 Peptide
  • Protein/Peptide Name:
    LyP-1, Peptide 1
  • Purity:
    95%
  • Molecular Weight:
    992.2 Da
  • Sequence:
    CGNKRTRGC (S-S Bonded)
  • Cat. No.:
    103007-222
  • Supplier no.:
    AS-62169

Specifications

About this item

LyP-1 recognizes lymphatics and tumor cells in certain tumors, but not lymphatics in normal tissues. Screening on breast carcinoma xenografts shows positive to cyclic 9-amino-acid peptide, LyP-1. The LyP-1 also recognizes an osteosarcoma xenograft, and spontaneous prostate and breast cancers in transgenic mice. LyP-1 peptide is detected in tumor structures that are positive for several lymphatic endothelial markers and negative for blood vessel markers. LyP-1 accumulates in the nuclei of the putative lymphatic cells, and in the nuclei of tumor cells.
Sequence:CGNKRTRGC (S-S Bonded)
MW:992.2 Da
% peak area by HPLC:95
Storage condition:-20° C