Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse;Rat Kisspeptin-10 (Kp-10)
Mouse;Rat Kisspeptin-10 (Kp-10)
Catalog # 103007-822
Supplier:  Anaspec Inc
CAS Number:  
Mouse;Rat Kisspeptin-10 (Kp-10)
Catalog # 103007-822
Supplier:  Anaspec Inc
Supplier Number:  AS-64240
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Rat
    Mouse
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Metastin Peptide
  • Protein/Peptide Name:
    Kisspeptin-10 (Kp-10)
  • Purity:
    95%
  • Molecular Weight:
    1302.5 Da
  • Sequence:
    YNWNSFGLRF-NH2
  • Cat. No.:
    103007-822
  • Supplier no.:
    AS-64240

Specifications

About this item

Kisspeptin was originally identified as a human metastasis suppressor gene that has the ability to suppress melanoma and breast cancer metastasis. Kisspeptin-GPR54 signaling has an important role in initiating secretion of gonadotropin-releasing hormone (GnRH) at puberty from the anterior pituitary.
This peptide sequence is found in residues 45 to 54 of Metastin (also referred to as Kisspeptin-10). This peptide increases plasma concentrations of GH (Growth Hormone) and LH (Luteinizing Hormone) in prepubertal dairy heifers. This peptide is the minimal sequence needed to activate GPR54 signaling, which may function in negatively regulating CXCR4’s role in programming tumor metastasis. Specifically, Kisspeptin-10 inhibits signaling and chemotaxis induced by SDF-1.
Sequence:YNWNSFGLRF-NH2
MW:1302.5 Da
% peak area by HPLC:95
Storage condition:-20° C