Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:p53N|Mdm-2 binding P53 domain
- Protein/Peptide Name:p53 (17-26)
- Purity:95%
- Molecular Weight:1753.1 Da
- Sequence:FITC-LC-ETFSDLWKLL-NH2
- Cat. No.:103007-320
- Supplier no.:AS-62386
This is amino acids 17 to 26 fragment of p53, fluorescent labeled through an LC spacer. This peptide is the Mdm-2 binding domain of p53 known also as p53N. This sequence contains all of the residues that come into contact with the binding domain of Mdm-2. The tumor suppressor protein p53 is important in maintaining genome stability and in preventing cancer development, FITC (Abs/Em=493 nm/517 nm)..
Sequence:FITC-LC-ETFSDLWKLL-NH2
MW:1753.1 Da
% peak area by HPLC:95
Storage condition:-20° C