Order Entry
Puerto Rico
ContactUsLinkComponent
 
Bak BH3 peptide, TAMRA (5-Carboxytetramethylrhodamin)
Catalog # 103008-254
Supplier:  Anaspec Inc
CAS Number:  
Bak BH3 peptide, TAMRA (5-Carboxytetramethylrhodamin)
Catalog # 103008-254
Supplier:  Anaspec Inc
Supplier Number:  AS-64590
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    TAMRA (5-Carboxytetramethylrhodamin)
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Flu-BakBH3 Peptide|BH3 domain of Bak Peptide
  • Protein/Peptide Name:
    Bak BH3 Peptide
  • Purity:
    95%
  • Molecular Weight:
    2137.4 Da
  • Sequence:
    5-TAMRA-GQVGRQLAIIGDDINR
  • Cat. No.:
    103008-254
  • Supplier no.:
    AS-64590

Specifications

About this item

This peptide is derived from the BH3 domain of Bak (Flu-BakBH3). It has high-affinity binding to a surface pocket of the Bcl-XL protein that is essential for its death antagonist function. This peptide is labeled with 5-TAMRA on the N-terminus, Abs/Em= 541/568
Sequence:5-TAMRA-GQVGRQLAIIGDDINR
MW:2137.4 Da
% peak area by HPLC:95
Storage condition:-20° C