CKS-17 (dimer)
A dimer of CKS-17 has a natural occurring cysteine at the carboxyl terminus (where it occurs in the retroviral peptides on which CKS-17 is based) and dimerization is accomplished by cysteine-disulfide linkage. CKS-17 is a synthetic retroviral envelope heptadecapeptide corresponding to a region highly conserved in retroviral transmembrane proteins such as pl5E. The CKS-17 peptide has been previously shown to inhibit monocyte superoxide production, natural killer cell activity, polyclonal B-cell activation, and monocyte-mediated killing by inactivation of interleukin-1.
Sequence:LQNRRGLDLLFLKEGGLC (dimer)
MW:4088.8 Da
% peak area by HPLC:95
Storage condition:-20° C
- Size:1 mg
- Storage Conditions:–20 °C
- Cat. No.:103008-252
- Molecular Weight:4088.8 Da
- Purity:95%
- Protein Synonyms:synthetic retroviral envelope heptadecapeptide|CSK 17 Peptide
- Protein/Peptide Name:CKS-17 (dimer)
- Conjugation:Unconjugated
- Sequence:LQNRRGLDLLFLKEGGLC (dimer)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



