Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:H-2Db restricted LCMV glycoprotein GP61
- Protein/Peptide Name:Lymphocytic Choreomeningitis Virus (LCMV) Glycoprotein 61
- Purity:95%
- Molecular Weight:2276.5 Da
- Sequence:GLNGPDIYKGVYQFKSVEFD
- Cat. No.:103008-236
- Supplier no.:AS-64560
An H-2Db restricted epitope, this peptide is amino acids 61 to 80 fragment of the lymphocytic choriomeningitis virus (LCMV) pre-glycoprotein polyprotein GP complex. LCMV has been routinely used for the study of adaptive immune responses to viral infection.
Sequence:GLNGPDIYKGVYQFKSVEFD
MW:2276.5 Da
% peak area by HPLC:95
Storage condition:-20° C