Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:LCMV nucleoprotein fragment|rat insulin promoter Lymphocytic Choriomeningitis Virus–Nucleoprotein fragment
- Protein/Peptide Name:LCMV NP396 H-2Db Peptide
- Purity:95%
- Molecular Weight:1078.2 Da
- Sequence:FQPQNGQFI
- Cat. No.:103006-908
- Supplier no.:AS-61700
This peptide is a rat insulin promoter Lymphocytic Choriomeningitis Virus–Nucleoprotein (LCMV-NP) fragment amino acid residues 396 to 404. It is the immunodominant H-2Db restricted epitope.
Sequence:FQPQNGQFI
MW:1078.2 Da
% peak area by HPLC:95
Storage condition:-20° C