Order Entry
Puerto Rico
ContactUsLinkComponent
T20
T20
Catalog # 103006-446
Supplier:  Anaspec Inc
CAS Number:  
T20
Catalog # 103006-446
Supplier:  Anaspec Inc
Supplier Number:  AS-61235
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    gp41 mimetic Peptide
  • Protein/Peptide Name:
    T20
  • Purity:
    95%
  • Molecular Weight:
    4492 Da
  • Sequence:
    Ac-YTSLIHSLIEESQNQQEKNEQELLELDKWASLWNWF-NH2
  • Cat. No.:
    103006-446
  • Supplier no.:
    AS-61235

Specifications

About this item

This 36-residue synthetic peptide strongly inhibits HIV-1 viral fusion with CD4 cells with an EC50 of 1 ng/ml.
It is a peptide mimetic of an essential region within the viral envelope glycoprotein gp41 that functions by blocking gp41 structural rearrangements at a transitional pre-fusion conformation.
Sequence:Ac-YTSLIHSLIEESQNQQEKNEQELLELDKWASLWNWF-NH2
MW:4492 Da
% peak area by HPLC:95
Storage condition:-20° C