Order Entry
Puerto Rico
ContactUsLinkComponent
HIV Substrate, QXL® 520-HiLyte Fluor™ 488
HIV Substrate, QXL® 520-HiLyte Fluor™ 488
Catalog # 103003-284
Supplier:  Anaspec Inc
CAS Number:  
HIV Substrate, QXL® 520-HiLyte Fluor™ 488
Catalog # 103003-284
Supplier:  Anaspec Inc
Supplier Number:  AS-60635
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    QXL® 520-HiLyte Fluor™ 488
  • Size:
    0.1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Fluorogenic HIV-1 protease substrate
  • Protein/Peptide Name:
    HIV Substrate
  • Purity:
    95%
  • Molecular Weight:
    2008.1 Da
  • Sequence:
    QXL™520-GABA-SQNYPIVQ-K(HiLyte Fluor™ 488)-NH2
  • Cat. No.:
    103003-284
  • Supplier no.:
    AS-60635

Specifications

About this item

This fluorogenic HIV-1 protease substrate contains the FRET pair HiLyte488 (fluorophore) and QXL520 (quencher), and a γ-Abu spacer. This FRET substrate is cleaved by the HIV-1 protease to generate fuorescence (Ex = 490nm; Em = 520nm).
Sequence:QXL™520-GABA-SQNYPIVQ-K(HiLyte Fluor™ 488)-NH2
MW:2008.1 Da
% peak area by HPLC:95
Storage condition:-20° C