Order Entry
Puerto Rico
ContactUsLinkComponent
HIV Protease FRET Substrate I, DABCYL-EDANS
HIV Protease FRET Substrate I, DABCYL-EDANS
Catalog # 102996-366
Supplier:  Anaspec Inc
CAS Number:  
HIV Protease FRET Substrate I, DABCYL-EDANS
Catalog # 102996-366
Supplier:  Anaspec Inc
Supplier Number:  AS-22992
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    DABCYL-EDANS
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    HIV protease substrate
  • Protein/Peptide Name:
    HIV Protease FRET Substrate I
  • Purity:
    95%
  • Molecular Weight:
    1532.5 Da
  • Sequence:
    DABCYL-GABA-SQNYPIVQ-EDANS
  • Cat. No.:
    102996-366
  • Supplier no.:
    AS-22992

Specifications

About this item

DABCYL-GABA-Ser-Gln-Asn-Tyr-Pro-Ile-Val-Gln-EDANS is also called HIV protease substrate I in some literature. It is widely used for the continuous assay for HIV protease activity. The 11-kD protease (PR) encoded by the human immunodeficiency virus 1 (HIV-1) is essential for the correct processing of viral polyproteins and the maturation of infectious virus, and is therefore a target for the design of selective acquired immunodeficiency syndrome (AIDS) therapeutics. The FRET-based fluorogenic substrate is derived from a natural processing site for HIV-1 PR. Incubation of recombinant HIV-1 PR with the fluorogenic substrate resulted in specific cleavage at the Tyr-Pro bond and a time-dependent increase in fluorescence intensity that is linearly related to the extent of substrate hydrolysis. The fluorescence quantum yields of the HIV-1 PR substrate in the FRET assay increased by 40.0- and 34.4-fold, respectively, per mole of substrate cleaved. Because of its simplicity and precision in the determination of reaction rates required for kinetic analysis, this substrate offers many advantages over the commonly used HPLC or electrophoresis-based assays for peptide substrate hydrolysis by retroviral PRs. Abs/Em = 340nm/490nm.
Sequence:DABCYL-GABA-SQNYPIVQ-EDANS
MW:1532.5 Da
% peak area by HPLC:95
Storage condition:-20° C