Order Entry
Puerto Rico
ContactUsLinkComponent
Staphylococcus Staphylococcal Enterotoxin B Domain (SEB) (144-153)
Staphylococcus Staphylococcal Enterotoxin B Domain (SEB) (144-153)
Catalog # 103007-764
Supplier:  Anaspec Inc
CAS Number:  
Staphylococcus Staphylococcal Enterotoxin B Domain (SEB) (144-153)
Catalog # 103007-764
Supplier:  Anaspec Inc
Supplier Number:  AS-63835
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Staphylococcus
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Enterotoxin B domain Peptide
  • Protein/Peptide Name:
    Staphylococcal Enterotoxin B Domain (SEB) (144-153)
  • Purity:
    95%
  • Molecular Weight:
    1159.4 Da
  • Sequence:
    KKKVTAQELD
  • Cat. No.:
    103007-764
  • Supplier no.:
    AS-63835

Specifications

About this item

This peptide is Staphylococcal Enterotoxin B domain (SEB) amino acid residue 163-172. This peptide sequence is highly conserved. It has been shown to inhibit transcytosis of multiple staphylococcal enterotoxins, SEA, SEE, and TSST-1.
Sequence:KKKVTAQELD
MW:1159.4 Da
% peak area by HPLC:95
Storage condition:-20° C